Learn More
Abnova™ Human NEK8 Partial ORF (NP_835464, 482 a.a. - 581 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00284086-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the serine/threionine protein kinase family related to NIMA (never in mitosis, gene A) of Aspergillus nidulans. The encoded protein may play a role in cell cycle progression from G2 to M phase. Mutations in the related mouse gene are associated with a disease phenotype that closely parallels the juvenile autosomal recessive form of polycystic kidney disease in humans. [provided by RefSeq]
Sequence: SHSCPQQVPMPPGQEAQRVVCGIDSSMILTVPGQALACGSNRFNKLGLDHLSLGEEPVPHQQVEEALSFTLLGSAPLDQEPLLSIDLGTAHSAAVTASGDSpecifications
NP_835464 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SHSCPQQVPMPPGQEAQRVVCGIDSSMILTVPGQALACGSNRFNKLGLDHLSLGEEPVPHQQVEEALSFTLLGSAPLDQEPLLSIDLGTAHSAAVTASGD | |
RUO | |
NEK8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
284086 | |
NEK8 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
JCK/MGC138445/NEK12A | |
NEK8 | |
Recombinant | |
wheat germ expression system |