missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NEK7 Partial ORF (NP_598001, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00140609-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16111947

  • 335.00€ / 10µg
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

NIMA-related kinases share high amino acid sequence identity with the gene product of the Aspergillus nidulans 'never in mitosis A' gene, which controls initiation of mitosis.[supplied by OMIM]

Sequence: MDEQSQGMQGPPVPQFQPQKALRPDMGYNTLANFRIEKKIGRGQFSEVYRAACLLDGVPVALKKVQIFDLMDAKARADCIKEIDLLKQLNHPNVIKYYAS
Specifications

Specifications

NP_598001
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MDEQSQGMQGPPVPQFQPQKALRPDMGYNTLANFRIEKKIGRGQFSEVYRAACLLDGVPVALKKVQIFDLMDAKARADCIKEIDLLKQLNHPNVIKYYAS
RUO
NEK7
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
140609
NEK7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NEK7
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers