missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NEK4 Partial ORF (AAH63044, 680 a.a. - 781 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006787-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16126035

  • 545.00€ / 25µg

Estimated Shipment: 04-10-2022
to see stock.
Add to basket

Description

Description

Sequence: DVPVANPVSEFKLHRKYRDTLILHGKVAEEAEEIHFKELPSAIMPGSEKIRRLVEVLRTDVIRGLGVQLLEQVYDLLEEEDEFDREVSVSLTVSRCLCYRIF
Specifications

Specifications

AAH63044
Liquid
6787
NEK4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC33171/NRK2/STK2/pp12301
NEK4
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DVPVANPVSEFKLHRKYRDTLILHGKVAEEAEEIHFKELPSAIMPGSEKIRRLVEVLRTDVIRGLGVQLLEQVYDLLEEEDEFDREVSVSLTVSRCLCYRIF
RUO
NEK4
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers