Learn More
Abnova™ Human NEK11 Partial ORF (AAH28587, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079858-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the never in mitosis gene A family of kinases. The encoded protein localizes to the nucleoli, and may function with NEK2A in the S-phase checkpoint. The encoded protein appears to play roles in DNA replication and response to genotoxic stress. Alternatively spliced transcript variants have been described
Sequence: MLKFQEAAKCVSGSTAISTYPKTLIARRYVLQQKLGSGSFGTVYLVSDKKAKRGEELKVLKEISVGELNPNETVQANLEAQLLSKLDHPAIVKFHASFVESpecifications
AAH28587 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLKFQEAAKCVSGSTAISTYPKTLIARRYVLQQKLGSGSFGTVYLVSDKKAKRGEELKVLKEISVGELNPNETVQANLEAQLLSKLDHPAIVKFHASFVE | |
RUO | |
NEK11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
79858 | |
NEK11 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23495 | |
NEK11 | |
Recombinant | |
wheat germ expression system |