missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NDUFA9 Full-length ORF (NP_004993.1, 1 a.a. - 377 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004704-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100598

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MAAAAQSRVVRVLSMSRSAITAIATSVCHGPPCRQLHHALMPHGKGGRSSVSGIVATVFGATGFLGRYVVNHLGRMGSQVIIPYRCDKYDIMHLRPMGDLGQLLFLEWDARDKDSIRRVVQHSNVVINLIGRDWETKNFDFEDVFVKIPQAIAQLSKEAGVEKFIHVSHLNANIKSSSRYLRNKAVGEKVVRDAFPEAIIVKPSDIFGREDRFLNSFASMHRFGPIPLGSLGWKTVKQPVYVVDVSKGIVNAVKDPDANGKSFAFVGPSRYLLFHLVKYIFAVAHRLFLPFPLPLFAYRWVARVFEISPFEPWITRDKVERMHITDMKLPHLPGLEDLGIQATPLELKAIEVLRRHRTYRWLSAEIEDVKPAKTVNI
Specifications

Specifications

NP_004993.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC111043/NDUFS2L/SDR22E1
NDUFA9
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4704
NDUFA9 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAAAQSRVVRVLSMSRSAITAIATSVCHGPPCRQLHHALMPHGKGGRSSVSGIVATVFGATGFLGRYVVNHLGRMGSQVIIPYRCDKYDIMHLRPMGDLGQLLFLEWDARDKDSIRRVVQHSNVVINLIGRDWETKNFDFEDVFVKIPQAIAQLSKEAGVEKFIHVSHLNANIKSSSRYLRNKAVGEKVVRDAFPEAIIVKPSDIFGREDRFLNSFASMHRFGPIPLGSLGWKTVKQPVYVVDVSKGIVNAVKDPDANGKSFAFVGPSRYLLFHLVKYIFAVAHRLFLPFPLPLFAYRWVARVFEISPFEPWITRDKVERMHITDMKLPHLPGLEDLGIQATPLELKAIEVLRRHRTYRWLSAEIEDVKPAKTVNI
RUO
NDUFA9
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers