missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NDOR1 Full-length ORF (AAH15735.1, 1 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00027158-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16178612

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes an NADPH-dependent diflavin reductase that contains both flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) binding domains. The encoded protein is an enzyme that catalyzes the transfers electrons from NADPH through FAD and FMN cofactors to potential redox partners. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MPSPQLLVLFGSQTGTAQDVSERLGREARRRRLGCRVQALDSYPVVNLINEPLVIFVCATTGQGDPPDNMKNFWRFIFRKNLPSTALCQMDFAVLGLGDSSYAKFNFVAKKLHRRLLQLGGSALLPVCLGDDQHELGPDAAVDPWLRDLWDRVLGLYPPPPGLTEIPPGVPLPSKFTLLFLQEAPSTGSEGQRVAHPGSQEPPSESKPFLAPMISNQRVTGPSHFQDVRLIEFDILGSGISFAAGDVVLIQPSNSAAHVQRFCQVLGLDPDQLFMLQPREPDVSSPTRLPQPCSMRHLVSHYLDIASVPRRSFFELLACLSLHELEREKLLEFSSAQGQEELFEYCNRPRRTILEVLCDFPHTAAAIPPDYLLDLIPVIRPRAFSIASSLLTHPSRLQILVAVVQFQTRLKEPRRGLCSSWLASLDPGQGPVRVPLWVRPGSLAFPETPDTPVIMVGPGTGVAPFRAAIQERVAQGQTGNFLFFGCRWRDQDFYWEAEWQELEKRDCLTLIPAFSREQEQKIYVQHRLRELGSLVWELLDRQGAYFYLAGNAKSMPADVSEALMSIFQEEGGLCSPDAAAYLARLQQTRRFQTETWA
Specifications

Specifications

AAH15735.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
93.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC138148/NR1/bA350O14.9
NDOR1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
27158
NDOR1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPSPQLLVLFGSQTGTAQDVSERLGREARRRRLGCRVQALDSYPVVNLINEPLVIFVCATTGQGDPPDNMKNFWRFIFRKNLPSTALCQMDFAVLGLGDSSYAKFNFVAKKLHRRLLQLGGSALLPVCLGDDQHELGPDAAVDPWLRDLWDRVLGLYPPPPGLTEIPPGVPLPSKFTLLFLQEAPSTGSEGQRVAHPGSQEPPSESKPFLAPMISNQRVTGPSHFQDVRLIEFDILGSGISFAAGDVVLIQPSNSAAHVQRFCQVLGLDPDQLFMLQPREPDVSSPTRLPQPCSMRHLVSHYLDIASVPRRSFFELLACLSLHELEREKLLEFSSAQGQEELFEYCNRPRRTILEVLCDFPHTAAAIPPDYLLDLIPVIRPRAFSIASSLLTHPSRLQILVAVVQFQTRLKEPRRGLCSSWLASLDPGQGPVRVPLWVRPGSLAFPETPDTPVIMVGPGTGVAPFRAAIQERVAQGQTGNFLFFGCRWRDQDFYWEAEWQELEKRDCLTLIPAFSREQEQKIYVQHRLRELGSLVWELLDRQGAYFYLAGNAKSMPADVSEALMSIFQEEGGLCSPDAAAYLARLQQTRRFQTETWA
RUO
NDOR1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers