Learn More
Abnova™ Human NDN Full-length ORF (NP_002478.1, 1 a.a. - 321 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004692-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This intronless gene is located in the Prader-Willi syndrome deletion region. It is an imprinted gene and is expressed exclusively from the paternal allele. Studies in mouse suggest that the protein encoded by this gene may suppress growth in postmitotic neurons. [provided by RefSeq]
Sequence: MSEQSKDLSDPNFAAEAPNSEVHSSPGVSEGVPPSATLAEPQSPPLGPTAAPQAAPPPQAPNDEGDPKALQQAAEEGRAHQAPSAAQPGPAPPAPAQLVQKAHELMWYVLVKDQKKMIIWFPDMVKDVIGSYKKWCRSILRRTSLILARVFGLHLRLTSLHTMEFALVKALEPEELDRVALSNRMPMTGLLLMILSLIYVKGRGARESAVWNVLRILGLRPWKKHSTFGDVRKLITEEFVQMNYLKYQRVPYVEPPEYEFFWGSRASREITKMQIMEFLARVFKKDPQAWPSRYREALEEARALREANPTAHYPRSSVSEDSpecifications
NP_002478.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HsT16328/PWCR | |
NDN | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4692 | |
NDN (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSEQSKDLSDPNFAAEAPNSEVHSSPGVSEGVPPSATLAEPQSPPLGPTAAPQAAPPPQAPNDEGDPKALQQAAEEGRAHQAPSAAQPGPAPPAPAQLVQKAHELMWYVLVKDQKKMIIWFPDMVKDVIGSYKKWCRSILRRTSLILARVFGLHLRLTSLHTMEFALVKALEPEELDRVALSNRMPMTGLLLMILSLIYVKGRGARESAVWNVLRILGLRPWKKHSTFGDVRKLITEEFVQMNYLKYQRVPYVEPPEYEFFWGSRASREITKMQIMEFLARVFKKDPQAWPSRYREALEEARALREANPTAHYPRSSVSED | |
RUO | |
NDN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |