Learn More
Abnova™ Human ND1 Partial ORF (YP_003024026, 21 a.a. - 71 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand:  Abnova™ H00004535-Q01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16128011
  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
Description
Sequence: MLTERKILGYIQLRKGPNVVGPYGLLQPFADAIKLFTKEPLKPATSTITLY
Specifications
Specifications
YP_003024026
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.35kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MLTERKILGYIQLRKGPNVVGPYGLLQPFADAIKLFTKEPLKPATSTITLY
RUO
ND1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4535
ND1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MTND1
ND1
Recombinant
wheat germ expression system