missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NBEA Partial ORF (NP_056493.3, 1133 a.a. - 1220 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026960-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105606

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of a large, diverse group of A-kinase anchor proteins that target the activity of protein kinase A to specific subcellular sites by binding to its type II regulatory subunits. Brain-specific expression and coat protein-like membrane recruitment of a highly similar protein in mouse suggest an involvement in neuronal post-Golgi membrane traffic. Mutations in this gene may be associated with a form of autism. Alternatively spliced transcript variants may exist, but their full-length nature has not been determined. [provided by RefSeq]

Sequence: ADEKEDLPNSSTSFLFDKIPKQEEKLLPELSSNHIIPNIQDTQVHLGVSDDLGLLAHMTGSVDLTCTSSIIEEKEFKIHTTSDGMSSI
Specifications

Specifications

NP_056493.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ADEKEDLPNSSTSFLFDKIPKQEEKLLPELSSNHIIPNIQDTQVHLGVSDDLGLLAHMTGSVDLTCTSSIIEEKEFKIHTTSDGMSSI
RUO
NBEA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
26960
NBEA (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCL8B/LYST2
NBEA
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers