missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NARG1 Partial ORF (NP_476516.1, 764 a.a. - 862 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080155-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16170187

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase. [provided by RefSeq]

Sequence: HRLSAAKMVYYLDPSSQKRAIELATTLDESLTNRNLQTCMEVLEALYDGSLGDCKEAAEIYRANCHKLFPYALAFMPPGYEEDMKITVNGDSSAEAEEL
Specifications

Specifications

NP_476516.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HRLSAAKMVYYLDPSSQKRAIELATTLDESLTNRNLQTCMEVLEALYDGSLGDCKEAAEIYRANCHKLFPYALAFMPPGYEEDMKITVNGDSSAEAEEL
RUO
NARG1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
80155
NARG1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Ga19/NATH/TBDN100
NARG1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers