missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MYST3 Partial ORF (NP_006757, 81 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007994-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: ALPKPRNHGKLDNKQNVDWNKLIKRAVEGLAESGGSTLKSIERFLKGQKDVSALFGGSAASGFHQQLRLAIKRAIGHGRLLKDGPLYRLNTKATNVDGKSpecifications
NP_006757 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ALPKPRNHGKLDNKQNVDWNKLIKRAVEGLAESGGSTLKSIERFLKGQKDVSALFGGSAASGFHQQLRLAIKRAIGHGRLLKDGPLYRLNTKATNVDGK | |
RUO | |
MYST3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7994 | |
MYST3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KAT6A/MGC167033/MOZ/RUNXBP2/ZNF220 | |
MYST3 | |
Recombinant | |
wheat germ expression system |