missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYST2 Partial ORF (AAH32640, 512 a.a. - 611 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011143-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16196242

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Sequence: SDLGLISYRSYWKEVLLRYLHNFQGKEISIKEISQETAVNPVDIVSTLQALQMLKYWKGKHLVLKRQDLIDEWIAKEAKRSNSNKTMDPSCLKWTPPKGT
Specifications

Specifications

AAH32640
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SDLGLISYRSYWKEVLLRYLHNFQGKEISIKEISQETAVNPVDIVSTLQALQMLKYWKGKHLVLKRQDLIDEWIAKEAKRSNSNKTMDPSCLKWTPPKGT
RUO
MYST2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11143
MYST2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HBO1/HBOA/KAT7
MYST2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers