missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYBPC1 Partial ORF (NP_996556, 506 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004604-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101525

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

Sequence: DAYNVTLPAKVHVIDPPKIILDGLDADNTVTVIAGNKLRLEIPISGEPPPKAMWSRGDKAIMEGSGRIRTESYPDSSTLVIDIAERDDSGVYHINLKN
Specifications

Specifications

NP_996556
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DAYNVTLPAKVHVIDPPKIILDGLDADNTVTVIAGNKLRLEIPISGEPPPKAMWSRGDKAIMEGSGRIRTESYPDSSTLVIDIAERDDSGVYHINLKN
RUO
MYBPC1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4604
MYBPC1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MYBPCC/MYBPCS/slow-type
MYBPC1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers