Learn More
Abnova™ Human MXI1 Full-length ORF (NP_005953.4, 1 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004601-P02.25ug
Additional Details : Weight : 0.00010kg
Description
Expression of the c-myc gene, which produces an oncogenic transcription factor, is tightly regulated in normal cells but is frequently deregulated in human cancers. The protein encoded by this gene is a transcriptional repressor thought to negatively regulate MYC function, and is therefore a potential tumor suppressor. This protein inhibits the transcriptional activity of MYC by competing for MAX, another basic helix-loop-helix protein that binds to MYC and is required for its function. Defects in this gene are frequently found in patients with prostate tumors. Three alternatively spliced transcripts encoding different isoforms have been described. Additional alternatively spliced transcripts may exist but the products of these transcripts have not been verified experimentally. [provided by RefSeq]
Sequence: MERVKMINVQRLLEAAEFLERRERECEHGYASSFPSMPSPRLQHSKPPRRLSRAQKHSSGSSNTSTANRSTHNELEKNRRAHLRLCLERLKVLIPLGPDCTRHTTLGLLNKAKAHIKKLEEAERKSQHQLENLEREQRFLKWRLEQLQGPQEMERIRMDSIGSTISSDRSDSEREEIEVDVESTEFSHGEVDNISTTSISDIDDHSSLPSIGSDEGYSSASVKLSFTSSpecifications
NP_005953.4 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAD2/MGC43220/MXD2/MXI/bHLHc11 | |
MXI1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4601 | |
MXI1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MERVKMINVQRLLEAAEFLERRERECEHGYASSFPSMPSPRLQHSKPPRRLSRAQKHSSGSSNTSTANRSTHNELEKNRRAHLRLCLERLKVLIPLGPDCTRHTTLGLLNKAKAHIKKLEEAERKSQHQLENLEREQRFLKWRLEQLQGPQEMERIRMDSIGSTISSDRSDSEREEIEVDVESTEFSHGEVDNISTTSISDIDDHSSLPSIGSDEGYSSASVKLSFTS | |
RUO | |
MXI1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |