Learn More
Abnova™ Human MSN Partial ORF (NP_002435, 422 a.a. - 531 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004478-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Moesin (for membrane-organizing extension spike protein) is a member of the ERM family which includes ezrin and radixin. ERM proteins appear to function as cross-linkers between plasma membranes and actin-based cytoskeletons. Moesin is localized to filopodia and other membranous protrusions that are important for cell-cell recognition and signaling and for cell movement. [provided by RefSeq]
Sequence: AELTARISQLEMARQKKESEAVEWQQKAQMVQEDLEKTRAELKTAMSTPHVAEPAENEQDEQDENGAEASADLRADAMAKDRSEEERTTEAEKNERVQKHLKALTSELANSpecifications
NP_002435 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AELTARISQLEMARQKKESEAVEWQQKAQMVQEDLEKTRAELKTAMSTPHVAEPAENEQDEQDENGAEASADLRADAMAKDRSEEERTTEAEKNERVQKHLKALTSELAN | |
RUO | |
MSN | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4478 | |
MSN (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MSN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |