missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MSC Full-length ORF (NP_005089.2, 1 a.a. - 206 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009242-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193432

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a transcriptional repressor capable of binding an E-box element either as a homodimer or as a heterodimer with E2A in vitro. The encoded protein also forms heterodimers with E2A proteins in vivo. This protein is capable of inhibiting the transactivation capability of E47, an E2A protein, in mammalian cells. This gene is a downstream target of the B-cell receptor signal transduction pathway. [provided by RefSeq]

Sequence: MSTGSVSDPEEMELRGLQREYPVPASKRPPLRGVERSYASPSDNSSAEEEDPDGEEERCALGTAGSAEGCKRKRPRVAGGGGAGGSAGGGGKKPLPAKGSAAECKQSQRNAANARERARMRVLSKAFSRLKTSLPWVPPDTKLSKLDTLRLASSYIAHLRQLLQEDRYENGYVHPVNLTWPFVVSGRPDSDTKEVSAANRLCGTTA
Specifications

Specifications

NP_005089.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ABF-1/ABF1/MYOR/bHLHa22
MSC
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9242
MSC (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSTGSVSDPEEMELRGLQREYPVPASKRPPLRGVERSYASPSDNSSAEEEDPDGEEERCALGTAGSAEGCKRKRPRVAGGGGAGGSAGGGGKKPLPAKGSAAECKQSQRNAANARERARMRVLSKAFSRLKTSLPWVPPDTKLSKLDTLRLASSYIAHLRQLLQEDRYENGYVHPVNLTWPFVVSGRPDSDTKEVSAANRLCGTTA
RUO
MSC
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers