missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MS4A6E Full-length ORF (NP_640342.1, 1 a.a. - 147 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00245802-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16169543

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. The gene encoding this protein is localized to 11q12.3, among a cluster of family members. [provided by RefSeq]

Sequence: MTSQPISNETIIMLPSNVINFSQAEKPEPTNQGQDSLKKRLQAKVKVIGVHSSLAGSILSALSALVGFILLSVNPAALNPASLQCKLDEKDIPTRLLLSYDYHSPYTMDCHRAKASLAGTLSLMLVSTVLEFCLAVLTAVLQWKQTV
Specifications

Specifications

NP_640342.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
42.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126832
MS4A6E
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
245802
MS4A6E (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTSQPISNETIIMLPSNVINFSQAEKPEPTNQGQDSLKKRLQAKVKVIGVHSSLAGSILSALSALVGFILLSVNPAALNPASLQCKLDEKDIPTRLLLSYDYHSPYTMDCHRAKASLAGTLSLMLVSTVLEFCLAVLTAVLQWKQTV
RUO
MS4A6E
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers