Learn More
Abcam Human MRPL2 knockout HEK-293T cell line
Product Code. 32158017 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32158017 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32158017 Supplier Abcam Supplier No. ab2666982x1000000Cells_
 Restricted Item
Estimated Shipment: 09-12-2026
MRPL2 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 100 bp deletion in exon 1 and 43 bp insertion in exon 1 and 7 bp deletion in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cel...
MRPL2 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 100 bp deletion in exon 1 and 43 bp insertion in exon 1 and 7 bp deletion in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells