missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MRAP Full-length ORF (NP_848932.1, 1 a.a. - 172 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00056246-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MANGTNASAPYYSYEYYLDYLDLIPVDEKKLKAHKHSIVIAFWVSLAAFVVLLFLILLYMSWSASPQMRNSPKHHQTCPWSHGLNLHLCIQKCLPCHREPLATSQAQASSVEPGSRTGPDQPLRQESSSTLPLGGFQTHPTLLWELTLNGGPLVRSKPSEPPPGDRTSQLQSSpecifications
NP_848932.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B27/C21orf61/FALP/FGD2/GCCD2 | |
MRAP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
56246 | |
MRAP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MANGTNASAPYYSYEYYLDYLDLIPVDEKKLKAHKHSIVIAFWVSLAAFVVLLFLILLYMSWSASPQMRNSPKHHQTCPWSHGLNLHLCIQKCLPCHREPLATSQAQASSVEPGSRTGPDQPLRQESSSTLPLGGFQTHPTLLWELTLNGGPLVRSKPSEPPPGDRTSQLQS | |
RUO | |
MRAP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |