missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MPI Full-length ORF (NP_002426.1, 1 a.a. - 423 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004351-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16160288

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib. [provided by RefSeq]

Sequence: MAAPRVFPLSCAVQQYAWGKMGSNSEVARLLASSDPLAQIAEDKPYAELWMGTHPRGDAKILDNRISQKTLSQWIAENQDSLGSKVKDTFNGNLPFLFKVLSVETPLSIQAHPNKELAEKLHLQAPQHYPDANHKPEMAIALTPFQGLCGFRPVEEIVTFLKKVPEFQFLIGDEAATHLKQTMSHDSQAVASSLQSCFSHLMKSEKKVVVEQLNLLVKRISQQAAAGNNMEDIFGELLLQLHQQYPGDIGCFAIYFLNLLTLKPGEAMFLEANVPHAYLKGDCVECMACSDNTVRAGLTPKFIDVPTLCEMLSYTPSSSKDRLFLPTRSQEDPYLSIYDPPVPDFTIMKTEVPGSVTEYKVLALDSASILLMVQGTVIASTPTTQTPIPLQRGGVLFIGANESVSLKLTEPKDLLIFRACCLL
Specifications

Specifications

NP_002426.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ39201/PMI/PMI1
MPI
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4351
MPI (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAPRVFPLSCAVQQYAWGKMGSNSEVARLLASSDPLAQIAEDKPYAELWMGTHPRGDAKILDNRISQKTLSQWIAENQDSLGSKVKDTFNGNLPFLFKVLSVETPLSIQAHPNKELAEKLHLQAPQHYPDANHKPEMAIALTPFQGLCGFRPVEEIVTFLKKVPEFQFLIGDEAATHLKQTMSHDSQAVASSLQSCFSHLMKSEKKVVVEQLNLLVKRISQQAAAGNNMEDIFGELLLQLHQQYPGDIGCFAIYFLNLLTLKPGEAMFLEANVPHAYLKGDCVECMACSDNTVRAGLTPKFIDVPTLCEMLSYTPSSSKDRLFLPTRSQEDPYLSIYDPPVPDFTIMKTEVPGSVTEYKVLALDSASILLMVQGTVIASTPTTQTPIPLQRGGVLFIGANESVSLKLTEPKDLLIFRACCLL
RUO
MPI
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers