missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MPHOSPH9 Full-length ORF (NP_073619.2, 1 a.a. - 1031 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010198-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194622

  • 528.00€ / 25µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

Sequence: MGFFSLSSERNESVIHYPESTEPEIQQEMSTSQPDCNVDSCSVSSGYGTFCISELNLYKSKDPKEFMEHIDVPKGQYVAPAVPAESLVDGVKNENFYIQTPEECHVSLKEDVSISPGEFEHNFLGENKVSEVYSGKTNSNAITSWAQKLKQNQPKRAHVEDGGSRSKQGNEQSKKTPIEKSDFAAATHPRAFYLSKPDETPNAWMSDSGTGLTYWKLEEKDMHHSLPETLEKTFISLSSTDVSPNQSNTSNEMKLPSLKDIYYKKQRENKQLPERNLTSASNPNHPPEVLTLDPTLHMKPKQQISGIQPHGLPNALDDRISFSPDSVLEPSMSSPSDIDSFSQASNVTSQLPGFPKYPSHTKASPVDSWKNQTFQNESRTSSTFPSVYTITSNDISVNTVDEENTVMVASASVSQSQLPGTANSVPECISLTSLEDPVILSKIRQNLKEKHARHIADLRAYYESEINSLKQKLEAKEISGVEDWKITNQILVDRCGQLDSALHEATSRVRTLENKNNLLEIEVNDLRERFSAASSASKILQERIEEMRTSSKEKDNTIIRLKSRLQDLEEAFENAYKLSDDKEAQLKQENKMFQDLLGEYESLGKEHRRVKDALNTTENKLLDAYTQISDLKRMISKLEAQVKQVEHENMLSLRHNSRIHVRPSRANTLATSDVSRRKWLIPGAEYSIFTGQPLDTQDSNVDNQLEETCSLGHRSPLEKDSSPGSSSTSLLIKKQRETSDTPIMRALKELDEGKIFKNWGTQTEKEDTSNINPRQTETSVNASRSPEKCAQQRQKRLNSASQRSSSLPPSNRKSSTPTKREIMLTPVTVAYSPKRSPKENLSPGFSHLLSKNESSPIRFDILLDDLDTVPVSTLQRTNPRKQLQFLPLDDSEEKTYSEKATDNHVNHSSCPEPVPNGVKKVSVRTAWEKNKSVSYEQCKPVSVTPQGNDFEYTAKIRTLAETERFFDELTKEKDQIEAALSRMPSPGGRITLQTRLNQEALEDRLERINRELGSVRMTLKKFHVLRTSANL
Specifications

Specifications

NP_073619.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
142.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434J034/FLJ12954/MPP-9/MPP9
MPHOSPH9
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10198
MPHOSPH9 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGFFSLSSERNESVIHYPESTEPEIQQEMSTSQPDCNVDSCSVSSGYGTFCISELNLYKSKDPKEFMEHIDVPKGQYVAPAVPAESLVDGVKNENFYIQTPEECHVSLKEDVSISPGEFEHNFLGENKVSEVYSGKTNSNAITSWAQKLKQNQPKRAHVEDGGSRSKQGNEQSKKTPIEKSDFAAATHPRAFYLSKPDETPNAWMSDSGTGLTYWKLEEKDMHHSLPETLEKTFISLSSTDVSPNQSNTSNEMKLPSLKDIYYKKQRENKQLPERNLTSASNPNHPPEVLTLDPTLHMKPKQQISGIQPHGLPNALDDRISFSPDSVLEPSMSSPSDIDSFSQASNVTSQLPGFPKYPSHTKASPVDSWKNQTFQNESRTSSTFPSVYTITSNDISVNTVDEENTVMVASASVSQSQLPGTANSVPECISLTSLEDPVILSKIRQNLKEKHARHIADLRAYYESEINSLKQKLEAKEISGVEDWKITNQILVDRCGQLDSALHEATSRVRTLENKNNLLEIEVNDLRERFSAASSASKILQERIEEMRTSSKEKDNTIIRLKSRLQDLEEAFENAYKLSDDKEAQLKQENKMFQDLLGEYESLGKEHRRVKDALNTTENKLLDAYTQISDLKRMISKLEAQVKQVEHENMLSLRHNSRIHVRPSRANTLATSDVSRRKWLIPGAEYSIFTGQPLDTQDSNVDNQLEETCSLGHRSPLEKDSSPGSSSTSLLIKKQRETSDTPIMRALKELDEGKIFKNWGTQTEKEDTSNINPRQTETSVNASRSPEKCAQQRQKRLNSASQRSSSLPPSNRKSSTPTKREIMLTPVTVAYSPKRSPKENLSPGFSHLLSKNESSPIRFDILLDDLDTVPVSTLQRTNPRKQLQFLPLDDSEEKTYSEKATDNHVNHSSCPEPVPNGVKKVSVRTAWEKNKSVSYEQCKPVSVTPQGNDFEYTAKIRTLAETERFFDELTKEKDQIEAALSRMPSPGGRITLQTRLNQEA
RUO
MPHOSPH9
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers