Learn More
Abnova™ Human MOV10L1 Partial ORF (NP_061868, 436 a.a. - 545 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054456-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is similar to a mouse gene that encodes a putative RNA helicase and shows testis-specific expression. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: FLIGRYLEVNVISGEESLIAAREPFSWKKLKSSQALTSAKTTVVVTAQKRNSRRQLPSFLPQYPIPDRLRKCVEQKIDILTFQPLLAELLNMSNYKEKFSTLLWLEEIYASpecifications
NP_061868 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FLIGRYLEVNVISGEESLIAAREPFSWKKLKSSQALTSAKTTVVVTAQKRNSRRQLPSFLPQYPIPDRLRKCVEQKIDILTFQPLLAELLNMSNYKEKFSTLLWLEEIYA | |
RUO | |
MOV10L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54456 | |
MOV10L1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DJ402G11.8/DKFZp434B0717/FLJ33421 | |
MOV10L1 | |
Recombinant | |
wheat germ expression system |