missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human/Mouse/Rat Total GAPDH DuoSet IC ELISA
Description
The Human/Mouse/Rat Total GAPDH DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat GAPDH.
Specifications
Specifications
| Product Type | ELISA Development Kit for Intracellular Proteins |
| Quantity | 1 Kit |
| Species | Human, Mouse, Rat |
| Conjugate | Biotin |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Format | 2 96-well Plates |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 24383 |
| Synonym | aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, G3PDH, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?