missing translation for 'onlineSavingsMsg'
Learn More

Abcam Human/Mouse /Rat ERK2 ELISA Kit

Product Code. 32119220 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 384 Tests
1 x 96 Tests
10 x 96 Tests
Unit Size:
384 tests
96 tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119220 1 x 96 Tests 96 tests
32111988 10 x 96 Tests 96 tests
32119061 1 x 384 Tests 384 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32119220 Supplier Abcam Supplier No. ab3175501x96Tests
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human/Mouse /Rat ERK2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human/Mouse /Rat ERK2 with a sensitivity of 18.483 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step.

Human/Mouse /Rat ERK2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human/Mouse /Rat ERK2 with a sensitivity of 18.483 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step.

- Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
- Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Assay Range 78.125 to 5000 pg/mL
Assay Sensitivity 18.483 pg/mL
Content And Storage 4°C
Detectable Analytes Mapk1 (ERK2)
Assay Human/Mouse /Rat ERK2 ELISA Kit
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Sandwich ELISA
Sample Type Cell Lysates, Tissue Extracts
Sufficient For 96 Tests
For Use With (Application) ELISA
Gene Mapk1
Immunoassay Kit Grade RUO
Incubation Time 60 minutes
Measuring Range 78.125 to 5000 pg/mL
Protein Mapk1
Quantitation Range 78.125 to 5000 pg/mL
Quantity 1 x 96 Tests
Regulatory Status RUO
Research Discipline Oncology
Shipping Condition Blue Ice
Target Mapk1 (ERK2)
Target Kit Named Mapk1 (ERK2)
Test Time 90 minutes
Total Assay Time 90 minutes
Detection Limit 78.125 pg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.