missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human/Mouse/Rat Activin A Protein
CHO derived
Brand: R&D Systems™ 338-AC-500
This item is not returnable.
View return policy
Description
Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.2-1.2 ng/mL. The specific activity of Recombinant Human/Mouse/Rat Activin A is approximately 2.5 x 103 units/mg, which is calibrated against human Activin A WHO International Standard (NIBSC code: 91/626). Specific activity is for reference purposes only and is not routinely tested.Order Info
Ships at ambient temperature.
Specifications
| P08476 | |
| 3624 | |
| 500 µg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| <0.01 EU / 1 µg of the protein by the LAL method. | |
| 0.2 to 1.2ng/mL | |
| Unconjugated | |
| Reconstitute at 100-500 µg/mL in sterile 4 mM HCl. |
| Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | |
| M.W. (Observed): 14 kDa, reducing conditions 24 kDa, non-reducing conditions; M.W. (theoretical): 13 kDa (monomer) | |
| Chinese Hamster Ovary cell line, CHO-derived Activin A protein Gly311-Ser426 | |
| RUO | |
| INHBA | |
| Recombinant protein | |
| Yes |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction