missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MOGAT3 Partial ORF (NP_835470, 59 a.a. - 107 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00346606-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192857

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Acyl-CoA:monoacylglycerol acyltransferase (MOGAT; EC 2.3.1.22) catalyzes the synthesis of diacylglycerol from 2-monoacylglycerol and fatty acyl-CoA (Cheng et al., 2003 [PubMed 12618427]).[supplied by OMIM]

Sequence: YLVWLYVDWDTPNQGGRRSEWIRNRAIWRQLRDYYPVKLVKTAELPPDR
Specifications

Specifications

NP_835470
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.13kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YLVWLYVDWDTPNQGGRRSEWIRNRAIWRQLRDYYPVKLVKTAELPPDR
RUO
MOGAT3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
346606
MOGAT3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DC7/DGAT2L7/MGAT3/MGC119203/MGC119204
MOGAT3
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers