missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MOBKL2C Partial ORF (NP_958805, 121 a.a. - 216 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00148932-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102097

  • 335.00€ / 10µg

Estimated Shipment: 12-05-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is similar to the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: LMDWIEGLINDEEVFPTRVGVPFPKNFQQVCTKILTRLFRVFVHVYIHHFDSILSMGAEAHVNTCYKHFYYFIREFSLVDQRELEPLREMTERICH
Specifications

Specifications

NP_958805
Liquid
148932
MOBKL2C (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC26743/MOB3C
MOBKL2C
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LMDWIEGLINDEEVFPTRVGVPFPKNFQQVCTKILTRLFRVFVHVYIHHFDSILSMGAEAHVNTCYKHFYYFIREFSLVDQRELEPLREMTERICH
RUO
MOBKL2C
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers