Learn More
Abnova™ Human MOBKL2C Partial ORF (NP_958805, 121 a.a. - 216 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00148932-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is similar to the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: LMDWIEGLINDEEVFPTRVGVPFPKNFQQVCTKILTRLFRVFVHVYIHHFDSILSMGAEAHVNTCYKHFYYFIREFSLVDQRELEPLREMTERICHSpecifications
NP_958805 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LMDWIEGLINDEEVFPTRVGVPFPKNFQQVCTKILTRLFRVFVHVYIHHFDSILSMGAEAHVNTCYKHFYYFIREFSLVDQRELEPLREMTERICH | |
RUO | |
MOBKL2C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
148932 | |
MOBKL2C (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC26743/MOB3C | |
MOBKL2C | |
Recombinant | |
wheat germ expression system |