Learn More
Abnova™ Human MNDA Partial ORF (NP_002423.1, 311 a.a. - 407 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004332-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The myeloid cell nuclear differentiation antigen (MNDA) is detected only in nuclei of cells of the granulocyte-monocyte lineage. A 200-amino acid region of human MNDA is strikingly similar to a region in the proteins encoded by a family of interferon-inducible mouse genes, designated Ifi-201, Ifi-202, and Ifi-203, that are not regulated in a cell- or tissue-specific fashion. The 1.8-kb MNDA mRNA, which contains an interferon-stimulated response element in the 5-prime untranslated region, was significantly upregulated in human monocytes exposed to interferon alpha. MNDA is located within 2,200 kb of FCER1A, APCS, CRP, and SPTA1. In its pattern of expression and/or regulation, MNDA resembles IFI16, suggesting that these genes participate in blood cell-specific responses to interferons. [provided by RefSeq]
Sequence: QLYKQASGTMVYGLFMLQKKSVHKKNTIYEIQDNTGSMDVVGSGKWHNIKCEKGDKLRLFCLQLRTVDRKLKLVCGSHSFIKVIKAKKNKEGPMNVNSpecifications
NP_002423.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QLYKQASGTMVYGLFMLQKKSVHKKNTIYEIQDNTGSMDVVGSGKWHNIKCEKGDKLRLFCLQLRTVDRKLKLVCGSHSFIKVIKAKKNKEGPMNVN | |
RUO | |
MNDA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4332 | |
MNDA (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PYHIN3 | |
MNDA | |
Recombinant | |
wheat germ expression system |