missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MNAB Partial ORF (NP_055369, 748 a.a. - 847 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054542-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: YVNAVDSRWSSYGNEATSSAHYVERDRFIVTDLSGHRKHSSTGDLLSLELQQAKSNSLLLQREANALAMQQKWNSLDEGRHLTLNLLSKEIELRNGEVKKSpecifications
NP_055369 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YVNAVDSRWSSYGNEATSSAHYVERDRFIVTDLSGHRKHSSTGDLLSLELQQAKSNSLLLQREANALAMQQKWNSLDEGRHLTLNLLSKEIELRNGEVKK | |
RUO | |
RC3H2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54542 | |
MNAB (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20301/FLJ20713/MGC52176/MNAB/RNF164 | |
RC3H2 | |
Recombinant | |
wheat germ expression system |