missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MNAB Partial ORF (NP_055369, 748 a.a. - 847 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054542-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107606

  • 528.00€ / 25µg

Estimated Shipment: 12-07-2023
to see stock.
Add to basket

Description

Description

Sequence: YVNAVDSRWSSYGNEATSSAHYVERDRFIVTDLSGHRKHSSTGDLLSLELQQAKSNSLLLQREANALAMQQKWNSLDEGRHLTLNLLSKEIELRNGEVKK
Specifications

Specifications

NP_055369
Liquid
54542
MNAB (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20301/FLJ20713/MGC52176/MNAB/RNF164
RC3H2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YVNAVDSRWSSYGNEATSSAHYVERDRFIVTDLSGHRKHSSTGDLLSLELQQAKSNSLLLQREANALAMQQKWNSLDEGRHLTLNLLSKEIELRNGEVKK
RUO
RC3H2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers