Learn More
Abnova™ Human MLPH Full-length ORF (NP_077006.1, 1 a.a. - 600 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079083-P01.10ug
Description
This gene encodes a member of the exophilin subfamily of Rab effector proteins. The protein forms a ternary complex with the small Ras-related GTPase Rab27A in its GTP-bound form and the motor protein myosin Va. A similar protein complex in mouse functions to tether pigment-producing organelles called melanosomes to the actin cytoskeleton in melanocytes, and is required for visible pigmentation in the hair and skin. A mutation in this gene results in Griscelli syndrome type 3, which is characterized by a silver-gray hair color and abnormal pigment distribution in the hair shaft. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: MGKKLDLSKLTDEEAQHVLEVVQRDFDLRRKEEERLEALKGKIKKESSKRELLSDTAHLNETHCARCLQPYQLLVNSKRQCLECGLFTCKSCGRVHPEEQGWICDPCHLARVVKIGSLEWYYEHVKARFKRFGSAKVIRSLHGRLQGGAGPELISEERSGDSDQTDEDGEPGSEAQAQAQPFGSKKKRLLSVHDFDFEGDSDDSTQPQGHSLHLSSVPEARDSPQSLTDESCSEKAAPHKAEGLEEADTGASGCHSHPEEQPTSISPSRHGALAELCPPGGSHRMALGTAAALGSNVIRNEQLPLQYLADVDTSDEESIRAHVMASHHSKRRGRASSESQIFELNKHISAVECLLTYLENTVVPPLAKGLGAGVRTEADVEEEALRRKLEELTSNVSDQETSSEEEEAKDEKAEPNRDKSVGPLPQADPEVGTAAHQTNRQEKSPQDPGDPVQYNRTTDEELSELEDRVAVTASEVQQAESEVSDIESRIAALRAAGLTVKPSGKPRRKSNLPIFLPRVAGKLGKRPEDPNADPSSEAKAMAVPYLLRRKFSNSLKSQGKDDDSFDRKSVYRGSLTQRNPNARKGMASHTFAKPVVAHQSSpecifications
NP_077006.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
92.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC2771/MGC59733/SLAC2-A | |
MLPH | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
79083 | |
MLPH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGKKLDLSKLTDEEAQHVLEVVQRDFDLRRKEEERLEALKGKIKKESSKRELLSDTAHLNETHCARCLQPYQLLVNSKRQCLECGLFTCKSCGRVHPEEQGWICDPCHLARVVKIGSLEWYYEHVKARFKRFGSAKVIRSLHGRLQGGAGPELISEERSGDSDQTDEDGEPGSEAQAQAQPFGSKKKRLLSVHDFDFEGDSDDSTQPQGHSLHLSSVPEARDSPQSLTDESCSEKAAPHKAEGLEEADTGASGCHSHPEEQPTSISPSRHGALAELCPPGGSHRMALGTAAALGSNVIRNEQLPLQYLADVDTSDEESIRAHVMASHHSKRRGRASSESQIFELNKHISAVECLLTYLENTVVPPLAKGLGAGVRTEADVEEEALRRKLEELTSNVSDQETSSEEEEAKDEKAEPNRDKSVGPLPQADPEVGTAAHQTNRQEKSPQDPGDPVQYNRTTDEELSELEDRVAVTASEVQQAESEVSDIESRIAALRAAGLTVKPSGKPRRKSNLPIFLPRVAGKLGKRPEDPNADPSSEAKAMAVPYLLRRKFSNSLKSQGKDDDSFDRKSVYRGSLTQRNPNARKGMASHTFAKPVVAHQS | |
RUO | |
MLPH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |