Learn More
Abnova™ Human MLC1SA Partial ORF (NP_002466, 109 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00140465-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in both slow-twitch skeletal muscle and in nonmuscle tissue. [provided by RefSeq]
Sequence: LGNPKSDELKSRRVDFETFLPMLQAVAKNRGQGTYEDYLEGFRVFDKEGNGKVMGAELRHVLTTLGEKMTEEEVETVLAGHEDSNGCINYEAFLKHILSVSpecifications
NP_002466 | |
Liquid | |
140465 | |
MLC1SA (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MLC1SA | |
MYL6B | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LGNPKSDELKSRRVDFETFLPMLQAVAKNRGQGTYEDYLEGFRVFDKEGNGKVMGAELRHVLTTLGEKMTEEEVETVLAGHEDSNGCINYEAFLKHILSV | |
RUO | |
MYL6B | |
Wheat Germ (in vitro) | |
GST |