missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MKI67IP Full-length ORF (NP_115766.2, 1 a.a. - 293 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084365-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195863

  • 349.00€ / 10µg

Estimated Shipment: 03-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a protein that interacts with the forkhead-associated domain of the Ki-67 antigen. The encoded protein may bind RNA and may play a role in mitosis and cell cycle progression. Multiple pseudogenes exist on chromosomes 5, 10, 12, 15, and 19.[provided by RefSeq]

Sequence: MATFSGPAGPILSLNPQEDVEFQKEVAQVRKRITQRKKQEQLTPGVVYVRHLPNLLDETQIFSYFSQFGTVTRFRLSRSKRTGNSKGYAFVEFESEDVAKIVAETMNNYLFGERLLECHFMPPEKVHKELFKDWNIPFKQPSYQSVKRYNRNRTLTQKLRMEERFKKKERLLRKKLAKKGIDYDFPSLILQKTESISKTNRQTSTKGQVLRKKKKKVSGTLDTPEKTVDSQGPTPVCTPTFLERRKSQVAELNDDDKDDEIVFKQPISCVKEEIQETQTPTHSRKKRRRSSNQ
Specifications

Specifications

NP_115766.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NIFK/Nopp34
MKI67IP
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84365
MKI67IP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATFSGPAGPILSLNPQEDVEFQKEVAQVRKRITQRKKQEQLTPGVVYVRHLPNLLDETQIFSYFSQFGTVTRFRLSRSKRTGNSKGYAFVEFESEDVAKIVAETMNNYLFGERLLECHFMPPEKVHKELFKDWNIPFKQPSYQSVKRYNRNRTLTQKLRMEERFKKKERLLRKKLAKKGIDYDFPSLILQKTESISKTNRQTSTKGQVLRKKKKKVSGTLDTPEKTVDSQGPTPVCTPTFLERRKSQVAELNDDDKDDEIVFKQPISCVKEEIQETQTPTHSRKKRRRSSNQ
RUO
MKI67IP
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers