Learn More
Abnova™ Human MINPP1 Partial ORF (NP_004888.2, 391 a.a. - 487 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009562-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes multiple inositol polyphosphate phosphatase; an enzyme that removes 3-phosphate from inositol phosphate substrates. It is the only enzyme known to hydrolzye inositol pentakisphosphate and inositol hexakisphosphate. This enzyme also converts 2,3 bisphosphoglycerate (2,3-BPG) to 2-phosphoglycerate; an activity formerly thought to be exclusive to 2,3-BPG synthase/2-phosphatase (BPGM) in the Rapoport-Luebering shunt of the glycolytic pathway
Sequence: TAYNYKKQMHRKFRSGLIVPYASNLIFVLYHCENAKTPKEQFRVQMLLNEKVLPLAYSQETVSFYEDLKNHYKDILQSCQTSEECELARANSTSDELSpecifications
NP_004888.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TAYNYKKQMHRKFRSGLIVPYASNLIFVLYHCENAKTPKEQFRVQMLLNEKVLPLAYSQETVSFYEDLKNHYKDILQSCQTSEECELARANSTSDEL | |
RUO | |
MINPP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9562 | |
MINPP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564L2016/HIPER1/MINPP2/MIPP | |
MINPP1 | |
Recombinant | |
wheat germ expression system |