missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MGC42105 Partial ORF (NP_699192, 342 a.a. - 435 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00167359-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192287

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Sequence: KHLSETSTLKEEENEVKSTLEHLGITEEHIRNNQGRDARSSITGVYRIILHRVQRKKALESVPVMMLPDPKERDLKKGSRVYRGIRHTSKFCSI
Specifications

Specifications

NP_699192
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.08kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KHLSETSTLKEEENEVKSTLEHLGITEEHIRNNQGRDARSSITGVYRIILHRVQRKKALESVPVMMLPDPKERDLKKGSRVYRGIRHTSKFCSI
RUO
MGC42105
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
167359
MGC42105 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NIM1
MGC42105
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers