missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MGC42105 Partial ORF (NP_699192, 342 a.a. - 435 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00167359-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: KHLSETSTLKEEENEVKSTLEHLGITEEHIRNNQGRDARSSITGVYRIILHRVQRKKALESVPVMMLPDPKERDLKKGSRVYRGIRHTSKFCSISpecifications
NP_699192 | |
Liquid | |
167359 | |
MGC42105 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NIM1 | |
MGC42105 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KHLSETSTLKEEENEVKSTLEHLGITEEHIRNNQGRDARSSITGVYRIILHRVQRKKALESVPVMMLPDPKERDLKKGSRVYRGIRHTSKFCSI | |
RUO | |
MGC42105 | |
Wheat Germ (in vitro) | |
GST |