missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MGC21874 Partial ORF (XP_291105, 2 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00093624-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101277

  • 335.00€ / 10µg

Estimated Shipment: 11-05-2023
to see stock.
Add to basket

Description

Description

Sequence: AELGKKYCVYCLAEVSPLRFRCTECQDIELCPECFSAGAEIGHHRRYHGYQLVDGGRFTLWGPEAEGGWTSREEQLLLDAIEQFGFGNWEDMAAHVGASRTPQEVMEHY
Specifications

Specifications

XP_291105
Liquid
93624
MGC21874 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADA2(beta)/ADA2B/MGC21874
TADA2B
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AELGKKYCVYCLAEVSPLRFRCTECQDIELCPECFSAGAEIGHHRRYHGYQLVDGGRFTLWGPEAEGGWTSREEQLLLDAIEQFGFGNWEDMAAHVGASRTPQEVMEHY
RUO
TADA2B
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers