missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human MGAT2 (NP_002399) VersaClone cDNA
Description
- Also known as: GNT2; CDG2A; CDGS2; GNT-II; GLCNACTII
- MGAT2 is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex Nglycans.
- MGAT2 has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signalanchor domain, and a C-terminal catalytic domain.
- Mutations in MGAT2 may lead to carbohydratedeficient glycoprotein syndrome, type II.
- The MGAT2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | MGAT2 |
| Accession Number | NP_002399 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1357 bp |
| Gene ID (Entrez) | 4247 |
| Gene Alias | alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase, Beta-1,2-N-acetylglucosaminyltransferase II, CDG2A, EC 2.4.1.143, GlcNAc-T II, GLCNACTII, GNT2, GNT-IICDGS2, Mannoside acetylglucosaminyltransferase 2, mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II, UDP-N-acetylglucosamine:alpha-6-D-mannosidebeta-1,2-N-acetylglucosaminyltransferase II |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?