missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Metabolic Luminex™ Performance Premixed Kit

Product Code. 30001023
Change view
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
2 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001023 6 Plex Each
30001002 7 Plex Each
30000974 10 Plex Each
18730097 3 Plex Each
18702997 8 Plex Each
30000953 9 Plex Each
30000930 2 Plex Each
30000852 11 Plex Each
30000850 12 Plex Each
18712447 4 Plex Each
30000824 5 Plex Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 30001023 Supplier R&D Systems™ Supplier No. FCSTM1906

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 6 Plex
Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.