Learn More
Abnova™ Human MEF2A Partial ORF (AAH13437.1, 71 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004205-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The process of differentiation from mesodermal precursor cells to myoblasts has led to the discovery of a variety of tissue-specific factors that regulate muscle gene expression. The myogenic basic helix-loop-helix proteins, including myoD (MIM 159970), myogenin (MIM 159980), MYF5 (MIM 159990), and MRF4 (MIM 159991) are one class of identified factors. A second family of DNA binding regulatory proteins is the myocyte-specific enhancer factor-2 (MEF2) family. Each of these proteins binds to the MEF2 target DNA sequence present in the regulatory regions of many, if not all, muscle-specific genes. The MEF2 genes are members of the MADS gene family (named for the yeast mating type-specific transcription factor MCM1, the plant homeotic genes 'agamous' and 'deficiens' and the human serum response factor SRF (MIM 600589)), a family that also includes several homeotic genes and other transcription factors, all of which share a conserved DNA-binding domain.[supplied by OMIM]
Sequence: EYNEPHESRTNSDIVEALNKKEHRGCDSPDPDTSYVLTPHTEEKYKKINEEFDNMMRNHKIAPGLPPQNFSMSVTVPVTSPNALSYTNPGSSLVSPSLAASpecifications
AAH13437.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
EYNEPHESRTNSDIVEALNKKEHRGCDSPDPDTSYVLTPHTEEKYKKINEEFDNMMRNHKIAPGLPPQNFSMSVTVPVTSPNALSYTNPGSSLVSPSLAA | |
RUO | |
MEF2A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4205 | |
MEF2A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADCAD1/RSRFC4/RSRFC9 | |
MEF2A | |
Recombinant | |
wheat germ expression system |