missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MED9 Full-length ORF (NP_060489.1, 1 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055090-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121113

  • 545.00€ / 25µg

Estimated Shipment: 07-10-2022
to see stock.
Add to basket

Description

Description

The multiprotein Mediator complex is a coactivator required for activation of RNA polymerase II transcription by DNA bound transcription factors. The protein encoded by this gene is thought to be a subunit of the Mediator complex. This gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq]

Sequence: MASAGVAAGRQAEDVLPPTSDQPLPDTKPLPPPQPPPVPAPQPQQSPAPRPQSPARAREEENYSFLPLVHNIIKCMDKDSPEVHQDLNALKSKFQEMRKLISTMPGIHLSPEQQQQQLQSLREQVRTKNELLQKYKSLCMFEIPKE
Specifications

Specifications

NP_060489.1
Liquid
55090
MED9 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASAGVAAGRQAEDVLPPTSDQPLPDTKPLPPPQPPPVPAPQPQQSPAPRPQSPARAREEENYSFLPLVHNIIKCMDKDSPEVHQDLNALKSKFQEMRKLISTMPGIHLSPEQQQQQLQSLREQVRTKNELLQKYKSLCMFEIPKE
RUO
MED9
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
42.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10193/MED25/MGC138234
MED9
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers