missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MED26 Full-length ORF (NP_004822.2, 1 a.a. - 600 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009441-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123662

  • 335.00€ / 10µg
Estimated Shipment: 27-05-2024
to see stock.
Add to basket

Description

Description

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. [provided by RefSeq]

Sequence: MTAAPASPQQIRDRLLQAIDPQSNIRNMVAVLEVISSLEKYPITKEALEETRLGKLINDVRKKTKNEELAKRAKKLLRSWQKLIEPAHQHEAALRGLAGATGSANGGAHNCRPEVGAAGPPRSIHDLKSRNDLQRLPGQRLDRLGSRKRRGDQRDLGHPGPPPKVSKASHDPLVPNSSPLPTNGISGSPESFASSLDGSGHAGPEGSRLERDENDKHSGKIPVNAVRPHTSSPGLGKPPGPCLQPKASVLQQLDRVDETPGPPHPKGPPRCSFSPRNSRHEGSFARQQSLYAPKGSVPSPSPRPQALDATQVPSPLPLAQPSTPPVRRLELLPSAESPVCWLEQPESHQRLAGPGCKAGLSPAEPLLSRAGFSPDSSKADSDAASSGGSDSKKKKRYRPRDYTVNLDGQVAEAGVKPVRLKERKLTFDPMTRQIKPLTQKEPVRADSPVHMEQQSRTELDKQEAKASLQSPFEQTNWKELSRNEIIQSYLSRQSSLLSSSGAQTPGAHHFMSEYLKQEESTRQGARQLHVLVPQSPPTDLPGLTREVTQDDLDRIQASQWPGVNGCQDTQGNWYDWTQCISLDPHGDDGRLNILPYVCLD
Specifications

Specifications

NP_004822.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
91.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CRSP7/CRSP70
MED26
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9441
MED26 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTAAPASPQQIRDRLLQAIDPQSNIRNMVAVLEVISSLEKYPITKEALEETRLGKLINDVRKKTKNEELAKRAKKLLRSWQKLIEPAHQHEAALRGLAGATGSANGGAHNCRPEVGAAGPPRSIHDLKSRNDLQRLPGQRLDRLGSRKRRGDQRDLGHPGPPPKVSKASHDPLVPNSSPLPTNGISGSPESFASSLDGSGHAGPEGSRLERDENDKHSGKIPVNAVRPHTSSPGLGKPPGPCLQPKASVLQQLDRVDETPGPPHPKGPPRCSFSPRNSRHEGSFARQQSLYAPKGSVPSPSPRPQALDATQVPSPLPLAQPSTPPVRRLELLPSAESPVCWLEQPESHQRLAGPGCKAGLSPAEPLLSRAGFSPDSSKADSDAASSGGSDSKKKKRYRPRDYTVNLDGQVAEAGVKPVRLKERKLTFDPMTRQIKPLTQKEPVRADSPVHMEQQSRTELDKQEAKASLQSPFEQTNWKELSRNEIIQSYLSRQSSLLSSSGAQTPGAHHFMSEYLKQEESTRQGARQLHVLVPQSPPTDLPGLTREVTQDDLDRIQASQWPGVNGCQDTQGNWYDWTQCISLDPHGDDGRLNILPYVCLD
RUO
MED26
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers