missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MCF2L Partial ORF (AAH20208, 220 a.a. - 319 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023263-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107112

  • 528.00€ / 25µg

Estimated Shipment: 04-07-2023
to see stock.
Add to basket

Description

Description

Sequence: GTELAETELPNDVQSTSSVLCAHTEKKDKAKEDLRLALKEGHSVLESLRELQAEGSEPSVNQDQLDNQATVQRLLAQLNETEAAFDEFWAKHQQKLEQCL
Specifications

Specifications

AAH20208
Liquid
23263
MCF2L (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARHGEF14/DBS/FLJ12122/KIAA0362/OST
MCF2L
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GTELAETELPNDVQSTSSVLCAHTEKKDKAKEDLRLALKEGHSVLESLRELQAEGSEPSVNQDQLDNQATVQRLLAQLNETEAAFDEFWAKHQQKLEQCL
RUO
MCF2L
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers