missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human MCAM/CD146 DuoSet ELISA

Product Code. 30001730
Change view
Click to view available options
Format:
96-well Microplate
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
30001730 96-well Microplate Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001730 Supplier R&D Systems™ Supplier No. DY93205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human MCAM/CD146 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human MCAM/CD146 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human MCAM/CD146.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Solid Phase Sandwich ELISA
Species Human
Binding Target Melanoma Cell Adhesion Molecule
Assay Range 125 to 8000 pg/mL
Format 96-well Microplate
Gene ID (Entrez) 4162 (Human); 84004 (Mouse); 78967 (Rat)
Sample Type Cell Culture Supernatants, Serum, EDTA Plasma, Heparin Plasma
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.