missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MC4R Partial ORF (NP_005903.1, 1 a.a. - 43 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004160-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107651

  • 528.00€ / 25µg

Estimated Shipment: 17-07-2023
to see stock.
Add to basket

Description

Description

Sequence: MVNSTHRGMHTSLHLWNRSSYRLHSNASESLGKGYSDGGCYEQ
Specifications

Specifications

NP_005903.1
Liquid
4160
MC4R (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126851/MGC138197
MC4R
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
30.47kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MVNSTHRGMHTSLHLWNRSSYRLHSNASESLGKGYSDGGCYEQ
RUO
MC4R
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers