Learn More
Abnova™ Human MBD3 Full-length ORF (NP_003917.1, 1 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00053615-P01.10ug
Additional Details : Weight : 0.00010kg
Description
DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). However, unlike the other family members, MBD3 is not capable of binding to methylated DNA. The predicted MBD3 protein shares 71% and 94% identity with MBD2 (isoform 1) and mouse Mbd3. MBD3 is a subunit of the NuRD, a multisubunit complex containing nucleosome remodeling and histone deacetylase activities. MBD3 mediates the association of metastasis-associated protein 2 (MTA2) with the core histone deacetylase complex. [provided by RefSeq]
Sequence: MERKRWECPALPQGWEREEVPRRSGLSAGHRDVFYYSPSGKKFRSKPQLARYLGGSMDLSTFDFRTGKMLMSKMNKSRQRVRYDSSNQVKGKPDLNTALPVRQTASIFKQPVTKITNHPSNKVKSDPQKAVDQPRQLFWEKKLSGLNAFDIAEELVKTMDLPKGLQGVGPGCTDETLLSAIASALHTSTMPITGQLSAAVEKNPGVWLNTTQPLCKAFMVTDEDIRKQEELVQQVRKRLEEALMADMLAHVEELARDGEAPLDKACAEDDDEEDEEEEEEEPDPDPEMEHVSpecifications
NP_003917.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MBD3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
53615 | |
MBD3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MERKRWECPALPQGWEREEVPRRSGLSAGHRDVFYYSPSGKKFRSKPQLARYLGGSMDLSTFDFRTGKMLMSKMNKSRQRVRYDSSNQVKGKPDLNTALPVRQTASIFKQPVTKITNHPSNKVKSDPQKAVDQPRQLFWEKKLSGLNAFDIAEELVKTMDLPKGLQGVGPGCTDETLLSAIASALHTSTMPITGQLSAAVEKNPGVWLNTTQPLCKAFMVTDEDIRKQEELVQQVRKRLEEALMADMLAHVEELARDGEAPLDKACAEDDDEEDEEEEEEEPDPDPEMEHV | |
RUO | |
MBD3 | |
Recombinant | |
wheat germ expression system |