Learn More
Abnova™ Human MBD1 Partial ORF (NP_056671, 415 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004152-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). Each of these proteins, with the exception of MBD3, is capable of binding specifically to methylated DNA. MECP2, MBD1 and MBD2 can also repress transcription from methylated gene promoters. Five transcript variants of the MBD1 are generated by alternative splicing resulting in protein isoforms that contain one MBD domain, two to three cysteine-rich (CXXC) domains, and some differences in the COOH terminus. All five transcript variants repress transcription from methylated promoters; in addition, variants with three CXXC domains also repress unmethylated promoter activity. MBD1 and MBD2 map very close to each other on chromosome 18q21. [provided by RefSeq]
Sequence: HHLGPTLKPTLATRTAQPDHTQAPTKQEAGGGFVLPPPGTDLVFLREGASSPVQVPGPVAASTEALLQEAQCSGLSWVVALPQVKQEKADTQDESpecifications
NP_056671 | |
Liquid | |
4152 | |
MBD1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CXXC3/PCM1/RFT | |
MBD1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HHLGPTLKPTLATRTAQPDHTQAPTKQEAGGGFVLPPPGTDLVFLREGASSPVQVPGPVAASTEALLQEAQCSGLSWVVALPQVKQEKADTQDE | |
RUO | |
MBD1 | |
Wheat Germ (in vitro) | |
GST |