Learn More
Abnova™ Human MAPK6 Full-length ORF (NP_002739.1, 1 a.a. - 721 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005597-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases). MAP kinases also known as extracellular signal-regulated kinases (ERKs), are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals. This kinase is localized in the nucleus, and has been reported to be activated in fibroblasts upon treatment with serum or phorbol esters. [provided by RefSeq]
Sequence: MAEKFESLMNIHGFDLGSRYMDLKPLGCGGNGLVFSAVDNDCDKRVAIKKIVLTDPQSVKHALREIKIIRRLDHDNIVKVFEILGPSGSQLTDDVGSLTELNSVYIVQEYMETDLANVLEQGPLLEEHARLFMYQLLRGLKYIHSANVLHRDLKPANLFINTEDLVLKIGDFGLARIMDPHYSHKGHLSEGLVTKWYRSPRLLLSPNNYTKAIDMWAAGCIFAEMLTGKTLFAGAHELEQMQLILESIPVVHEEDRQELLSVIPVYIRNDMTEPHKPLTQLLPGISREALDFLEQILTFSPMDRLTAEEALSHPYMSIYSFPMDEPISSHPFHIEDEVDDILLMDETHSHIYNWERYHDCQFSEHDWPVHNNFDIDEVQLDPRALSDVTDEEEVQVDPRKYLDGDREKYLEDPAFDTNYSTEPCWQYSDHHENKYCDLECSHTCNYKTRSSSYLDNLVWRESEVNHYYEPKLIIDLSNWKEQSKEKSDKKGKSKCERNGLVKAQIALEEASQQLAGKEREKNQGFDFDSFIAGTIQLSSQHEPTDVVDKLNDLNSSVSQLELKSLISKSVSQEKQEKGMANLAQLEALYQSSWDSQFVSGGEDCFFINQFCEVRKDEQVEKENTYTSYLDKFFSRKEDTEMLETEPVEDGKLGERGHEEGFLNNSGEFLFNKQLESIGIPQFHSPVGSPLKSIQATLTPSAMKSSPQIPHQTYSSILKHLNSpecifications
NP_002739.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
109.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686F03189/ERK3/HsT17250/PRKM6/p97MAPK | |
MAPK6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5597 | |
MAPK6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEKFESLMNIHGFDLGSRYMDLKPLGCGGNGLVFSAVDNDCDKRVAIKKIVLTDPQSVKHALREIKIIRRLDHDNIVKVFEILGPSGSQLTDDVGSLTELNSVYIVQEYMETDLANVLEQGPLLEEHARLFMYQLLRGLKYIHSANVLHRDLKPANLFINTEDLVLKIGDFGLARIMDPHYSHKGHLSEGLVTKWYRSPRLLLSPNNYTKAIDMWAAGCIFAEMLTGKTLFAGAHELEQMQLILESIPVVHEEDRQELLSVIPVYIRNDMTEPHKPLTQLLPGISREALDFLEQILTFSPMDRLTAEEALSHPYMSIYSFPMDEPISSHPFHIEDEVDDILLMDETHSHIYNWERYHDCQFSEHDWPVHNNFDIDEVQLDPRALSDVTDEEEVQVDPRKYLDGDREKYLEDPAFDTNYSTEPCWQYSDHHENKYCDLECSHTCNYKTRSSSYLDNLVWRESEVNHYYEPKLIIDLSNWKEQSKEKSDKKGKSKCERNGLVKAQIALEEASQQLAGKEREKNQGFDFDSFIAGTIQLSSQHEPTDVVDKLNDLNSSVSQLELKSLISKSVSQEKQEKGMANLAQLEALYQSSWDSQFVSGGEDCFFINQFCEVRKDEQVEKENTYTSYLDKFFSRKEDTEMLETEPVEDGKLGERGHEEGFLNNSGEFLFNKQLESIGIPQFHSPVGSPLKSIQATLTPSAMKSSPQIPHQTYSSILKHLN | |
RUO | |
MAPK6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |