Learn More
Abcam Human MAP3K2 (MEKK2) knockout A549 cell line
Description
MAP3K2 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 7 and 4 bp deletion in exon 7. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Quantity | 1,000,000 Cells |
| Regulatory Status | RUO |